Order Entry
Slovakia
ContactUsLinkComponent
Glycine CAS:56-40-6 EC:200-272-2 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 20 g
Catalog # CMPLSB18150.20G
New Product
Glycine CAS:56-40-6 EC:200-272-2 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 20 g
Catalog # CMPLSB18150.20G