Order Entry
Slovakia
ContactUsLinkComponent
Breath alcohol testing device for professional use, compliant to EN15964, US NHTSA evidential, Australia AS3547 and many other national and international standards and regulations. Passive tests and tests with mouthpiece possible. Manufactured in Ger 1 * 1 items
Catalog # DRAE3706961
Supplier:  Dräger
New Product
Breath alcohol testing device for professional use, compliant to EN15964, US NHTSA evidential, Australia AS3547 and many other national and international standards and regulations. Passive tests and tests with mouthpiece possible. Manufactured in Ger 1 * 1 items
Catalog # DRAE3706961
Supplier:  Dräger