Order Entry
Slovakia
ContactUsLinkComponent
3 mm Ø, borosilicate glass D 263® M, thickness No. 1 (0.13-0.16 mm), CE marked, 100 pcs./two-part plastic box 1 * 1.000 items
Catalog # MARI0111999491
Supplier:  MARIENFELD
New Product
3 mm Ø, borosilicate glass D 263® M, thickness No. 1 (0.13-0.16 mm), CE marked, 100 pcs./two-part plastic box 1 * 1.000 items
Catalog # MARI0111999491
Supplier:  MARIENFELD