Claudin18.2 IHC 3B10 Kit 1 * 8 mL
Catalog # ACRBCAA-B002-7.5ML
Supplier: ACRO Biosystems
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: BIOTIUM
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF