0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Observation Tube 200mm Long OT-200(I) (SUS) 1 * 1 items
ATAGRF-72079
:
Atago
Observation Tube 200mm Long OT-200(I) (SUS) 1 * 1 items
ATAGRF-72079
:
Atago
Frequently Bought Together
Potassium dihydrogen phosphate 1/15 mol/l in aqueous solution, Supelco®
: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
pH buffer solutions
: METTLER TOLEDO
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
: HAMILTON BONADUZ
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals