Order Entry
Slovakia
ContactUsLinkComponent
11 components: 100mg/l each of Ca Mg Na K Mn Fe Si P Cu Mo Zn in HNO3 2% 1 * 2 Bottle
Catalog # CMPLRM026377L1.2
11 components: 100mg/l each of Ca Mg Na K Mn Fe Si P Cu Mo Zn in HNO3 2% 1 * 2 Bottle
Catalog # CMPLRM026377L1.2
Frequently Bought Together