DISINFECTANT, INCIDIN OXYFOAM, 6X750ML 1 * 6 KS
Catalog # SHIE3115830
Supplier: ECOLAB CONTAMINATION CONTROL
DISINFECTANT, INCIDIN OXYFOAM, 6X750ML 1 * 6 KS
Catalog # SHIE3115830
Supplier: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: HONEYWELL
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Phenetole ≥98%
Supplier: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99%, pure
Supplier: Thermo Fisher Scientific
Methanol 99.9% for analysis
Supplier: Thermo Fisher Scientific
Methanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: HONEYWELL