0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
7 COMPONENT 0.5MG/L EACH IN H2O 1 * 100 mL
CMPLRM096100L1
:
CUSTOM MADE CHEMICALS LAB
7 COMPONENT 0.5MG/L EACH IN H2O 1 * 100 mL
CMPLRM096100L1
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
EOSIN RAL 555 1L 1 * 1 L
: RAL
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Formalin 10%, Q Path®, buffered
: VWR Chemicals
HISTOPLAS PLUS 8X 1KG 1 * 8 items
: SHANDON
HACH 100 ML SULFIDE 2 (100 TESTS) 1 * 100 mL
: WATER ID PRIMELAB
PHOSPHATE REAGENT I 1 * 250 mL
: TINTOMETER