KLERCIDE SPORICIDAL ENHANCED PEROXIDE 1L 1 * 6 items
Catalog # SHIE6600520
Supplier: ECOLAB CONTAMINATION CONTROL
KLERCIDE SPORICIDAL ENHANCED PEROXIDE 1L 1 * 6 items
Catalog # SHIE6600520
Supplier: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chemical resistant gloves, AlphaTec® Solvex 37-675
Supplier: Ansell
VWR®, Cover Glasses, Square
Supplier: VWR Collection
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals