RAC METANEPHRINE-D3 HYDROCHLORIDE 1 * 5 mg
Catalog # PROOTRCM258762-5MG
Supplier: PROMOCHEM
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: HACH
Lead acetate solution Reag. Ph. Eur. 1048103
Supplier: VWR Chemicals
pH buffer, single-use, SINGLET™
Supplier: HACH
TRICLABENDAZOLE FOR SYSTEM SUITABILITY 1 * 0,12 mg
Supplier: EDQM