PBDE 100, 50 µG/ML IN NONANE 1 * 1 mL
Catalog # EHERA15898100NO-50
Supplier: EHRENSTORFER
Frequently Bought Together
VWR®, Bromide 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Colour Reference solution, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
PIROCTONE OLAMINE 1 * 500 mg
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
VWR®, Colour standard solutions ASTM
Supplier: VWR Chemicals
Reagent for sulphate determination using Lovibond® MD photometers
Supplier: TINTOMETER
Estradiol
Supplier: EHRENSTORFER
Methyl Palmitate, Reference Standard
Supplier: USP