Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Bromonitromethane 90%

Bromonitromethane 90%

Bottle, 1 g
Catalog # APOSOR3067-1G
Oxidizing
Irritant
Supplier:   Apollo Scientific
Bromonitromethane 90%
Catalog # APOSOR3067-1G
Supplier:  Apollo Scientific
CAS Number:  563-70-2
Bromonitromethane 90%

Bottle, 1 g

CAS Number:  563-70-2
Price
33,82 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:563-70-2
  • Formula:BrCH₂NO₂
  • MDL Number:MFCD00007401
  • Molecular Weight:139.94 g/mol
  • Boiling Pt:146…148 °C/750mmHg
  • Density:2.007 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
  • EINECS:209-258-0
  • UN:3139
  • ADR:5.1,II
  •  :Density: 2.007 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications