STŘÍBRO DRÁT, ? 0,5 MM, VÁHA NA METR CCA 2,062 G 1 * 25 g
Catalog # ROTHR.8821.1
Supplier: ROTH CARL
STŘÍBRO DRÁT, ? 0,5 MM, VÁHA NA METR CCA 2,062 G 1 * 25 g
Catalog # ROTHR.8821.1
Supplier: ROTH CARL
Frequently Bought Together
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethyl acetate, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck