Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: SI. Inner Diameter (ID): 0.94 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary 1 * 1 items
Catalog # HEWL5500-1446
Supplier: AGILENT HEWLETT PACKARD
Frequently Bought Together