Order Entry
Slovakia
ContactUsLinkComponent
Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA2 (AM130) is a chimeric monoclonal antibody combining the constant domain s of the human IgA2 molecule with mouse variable regions. The variable region was obtained from a mouse immunized wit 1 * 1 mg
Catalog # ACRBSPD-M521-1MG
Supplier:  ACRO Biosystems
Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA2 (AM130) is a chimeric monoclonal antibody combining the constant domain s of the human IgA2 molecule with mouse variable regions. The variable region was obtained from a mouse immunized wit 1 * 1 mg
Catalog # ACRBSPD-M521-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together