Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Anti-SARS-CoV-2 Nucleocapsid Antibody, Chimeric mAb, Human IgG1 (AM223) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 const 1 * 10 mg
Catalog # ACRBNUN-BM272-10MG
Supplier:  ACRO Biosystems
Biotinylated Anti-SARS-CoV-2 Nucleocapsid Antibody, Chimeric mAb, Human IgG1 (AM223) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 const 1 * 10 mg
Catalog # ACRBNUN-BM272-10MG
Supplier:  ACRO Biosystems
Frequently Bought Together