Unconjugated,Anti-SARS-CoV-2 Omicron Antibody-3A7C12,Human,Human IgG1 | Human Kappa,HEK is a chimeric monoclonal antibody recombinan tly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 co 1 * 1 mg
Catalog # ACRBSPD-M415-1MG
Supplier: ACRO Biosystems
Frequently Bought Together