Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse SIRP alpha, His,Avitag (SIA-M82E2) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (A ccession # P97797-1). 1 * 200 µG
Catalog # ACRBSIA-M82E2200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse SIRP alpha, His,Avitag (SIA-M82E2) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (A ccession # P97797-1). 1 * 200 µG
Catalog # ACRBSIA-M82E2200UG
Supplier:  ACRO Biosystems
Frequently Bought Together