Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human SIRP gamma, His,Avitag (SIG-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Ser 364 (A ccession # AAH64532.1). 1 * 25 µG
Catalog # ACRBSIG-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human SIRP gamma, His,Avitag (SIG-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Ser 364 (A ccession # AAH64532.1). 1 * 25 µG
Catalog # ACRBSIG-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together