Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human GPRC5D, His,Avitag&Flag Tag (GPD-H82D6) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 - Phe 19 2 & Glu 197 - Leu 262 (Accession # Q9NZD1-2). 1 * 100 µG
Catalog # ACRBGPD-H82D6100UG
Supplier:  ACRO Biosystems
Biotinylated Human GPRC5D, His,Avitag&Flag Tag (GPD-H82D6) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 - Phe 19 2 & Glu 197 - Leu 262 (Accession # Q9NZD1-2). 1 * 100 µG
Catalog # ACRBGPD-H82D6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together