Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus GPRC5D, Flag,His Tag (Nanodisc) (GPD-C82D4) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 - Phe 192 & Glu 197 - Leu 262 (Accession # A0A2K5W6I7-1). 1 * 100 µG
Catalog # ACRBGPD-C82D4100UG
Supplier:  ACRO Biosystems
Cynomolgus GPRC5D, Flag,His Tag (Nanodisc) (GPD-C82D4) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 - Phe 192 & Glu 197 - Leu 262 (Accession # A0A2K5W6I7-1). 1 * 100 µG
Catalog # ACRBGPD-C82D4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together