Order Entry
Slovakia
ContactUsLinkComponent
Mouse GFR alpha-like, His Tag (GFA-M52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 349 (Accession # Q 6SJE0-1). 1 * 1 mg
Catalog # ACRBGFA-M52H7-1MG
Supplier:  ACRO Biosystems
Mouse GFR alpha-like, His Tag (GFA-M52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 349 (Accession # Q 6SJE0-1). 1 * 1 mg
Catalog # ACRBGFA-M52H7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together