Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human NKp46 Protein, His Tag, primary amine labeling (NK6-H82E2) is expressed from human 293 cells (HEK293). It contain s AA Gln 22 - Asn 254 (Accession # O76036-6). 1 * 200 µG
Catalog # ACRBNK6-H82E2200UG
Supplier:  ACRO Biosystems
Biotinylated Human NKp46 Protein, His Tag, primary amine labeling (NK6-H82E2) is expressed from human 293 cells (HEK293). It contain s AA Gln 22 - Asn 254 (Accession # O76036-6). 1 * 200 µG
Catalog # ACRBNK6-H82E2200UG
Supplier:  ACRO Biosystems
Frequently Bought Together