Order Entry
Slovakia
ContactUsLinkComponent
FITC-Labeled Human CEACAM-5, Fc Tag (CE5-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accessi on # P06731-1). It is the FITC labeled form of Human CEACAM-5, Fc Tag (Cat. No. CE5-H5253). 1 * 25 µG
Catalog # ACRBCE5-HF255-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human CEACAM-5, Fc Tag (CE5-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accessi on # P06731-1). It is the FITC labeled form of Human CEACAM-5, Fc Tag (Cat. No. CE5-H5253). 1 * 25 µG
Catalog # ACRBCE5-HF255-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together