Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus NKG2A&CD94, His Tag (NC4-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & As p 57 - Ile 179 (CD94) (Accession # Q68VD2-1 (NKG2A) & Q68VD4-1 (CD94)). 1 * 1 mg
Catalog # ACRBNC4-C52H6-1MG
Supplier:  ACRO Biosystems
Cynomolgus NKG2A&CD94, His Tag (NC4-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & As p 57 - Ile 179 (CD94) (Accession # Q68VD2-1 (NKG2A) & Q68VD4-1 (CD94)). 1 * 1 mg
Catalog # ACRBNC4-C52H6-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together