Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human APOE2, His,Avitag (APE-H52E5) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Access ion # P02649-1 (R176C)). 1 * 200 µG
Catalog # ACRBAPE-H52E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human APOE2, His,Avitag (APE-H52E5) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Access ion # P02649-1 (R176C)). 1 * 200 µG
Catalog # ACRBAPE-H52E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together