Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Rat TNFSF11, His,Avitag (RAL-R82Q9) is expressed from human 293 cells (HEK293). It contains AA Leu 69 - Asp 318 (Access ion # Q9ESE2-1). 1 * 200 µG
Catalog # ACRBRAL-R82Q9200UG
Supplier:  ACRO Biosystems
Biotinylated Rat TNFSF11, His,Avitag (RAL-R82Q9) is expressed from human 293 cells (HEK293). It contains AA Leu 69 - Asp 318 (Access ion # Q9ESE2-1). 1 * 200 µG
Catalog # ACRBRAL-R82Q9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together