Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human APOE3 (R154S), His,Avitag (APE-H52E4) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1 (R154S)). 1 * 25 µG
Catalog # ACRBAPE-H52E4-25UG
Supplier:  ACRO Biosystems
Biotinylated Human APOE3 (R154S), His,Avitag (APE-H52E4) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1 (R154S)). 1 * 25 µG
Catalog # ACRBAPE-H52E4-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together