Order Entry
Slovakia
ContactUsLinkComponent
Rat CD19, His Tag (CD9-R52H6) is expressed from human 293 cells (HEK293). It contains AA Arg 19 - Gly 287 (Accession # F1LNH2-1). 1 * 1 mg
Katalógové číslo ACRBCD9-R52H6-1MG
Supplier:  ACRO Biosystems
Rat CD19, His Tag (CD9-R52H6) is expressed from human 293 cells (HEK293). It contains AA Arg 19 - Gly 287 (Accession # F1LNH2-1). 1 * 1 mg
Katalógové číslo ACRBCD9-R52H6-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together