Biotinylated Human HLA-A*02:01 NY-ESO-1 (SLLMWITQC) Complex Protein (HL1-H82E6) is expressed from human 293 cells (HEK293). It conta ins AA Gly 25 - Ile 308 (HLA-A*0201) & SLLMWITQC & Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*0201) & CAA0 1 * 25 µG
Catalog # ACRBHL1-H82E6-25UG
Supplier: ACRO Biosystems
Frequently Bought Together