Order Entry
Slovakia
ContactUsLinkComponent
Human Neuropilin-2, His Tag (NR2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 21 - Pro 864 (Accession # O60 462-1). 1 * 1 mg
Catalog # ACRBNR2-H52H3-1MG
Supplier:  ACRO Biosystems
Human Neuropilin-2, His Tag (NR2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 21 - Pro 864 (Accession # O60 462-1). 1 * 1 mg
Catalog # ACRBNR2-H52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together