Order Entry
Slovakia
ContactUsLinkComponent
Canine CD36, His Tag (CD6-C52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # D5IGC7-1). 1 * 100 µG
Catalog # ACRBCD6-C52H6100UG
Supplier:  ACRO Biosystems
Canine CD36, His Tag (CD6-C52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # D5IGC7-1). 1 * 100 µG
Catalog # ACRBCD6-C52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together