Order Entry
Slovakia
ContactUsLinkComponent
Human LRRK2 (G2019S), His Tag (LK2-H5245) is expressed from human 293 cells (HEK293). It contains AA Met 968 - Gln 2527 (Accession # Q5S007 (G2019S)). 1 * 20 µG
Catalog # ACRBLK2-H5245-20UG
Supplier:  ACRO Biosystems
Human LRRK2 (G2019S), His Tag (LK2-H5245) is expressed from human 293 cells (HEK293). It contains AA Met 968 - Gln 2527 (Accession # Q5S007 (G2019S)). 1 * 20 µG
Catalog # ACRBLK2-H5245-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together