Order Entry
Slovakia
ContactUsLinkComponent
Human DDC, His Tag (DDC-H55H6) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Glu 480 (Accession # P20711-1). 1 * 20 µG
Catalog # ACRBDDC-H55H6-20UG
Supplier:  ACRO Biosystems
Human DDC, His Tag (DDC-H55H6) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Glu 480 (Accession # P20711-1). 1 * 20 µG
Catalog # ACRBDDC-H55H6-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together