Order Entry
Slovakia
ContactUsLinkComponent
Human GSK-3beta, His Tag (GSB-H5545) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Thr 420 (Accession # P49841- 1). 1 * 20 µG
Catalog # ACRBGSB-H5545-20UG
Supplier:  ACRO Biosystems
Human GSK-3beta, His Tag (GSB-H5545) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Thr 420 (Accession # P49841- 1). 1 * 20 µG
Catalog # ACRBGSB-H5545-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together