Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Fluorescent VLP isotype control (VLP-NF2P4) is expressed from human 293 cells (HEK293). 1 * 150 µG
Catalog # ACRBVLP-NF2P4150UG
Supplier:
ACRO Biosystems
Expand image
Fluorescent VLP isotype control (VLP-NF2P4) is expressed from human 293 cells (HEK293). 1 * 150 µG
Catalog # ACRBVLP-NF2P4150UG
Supplier:
ACRO Biosystems
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
VWR® Blender Bags
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection