Order Entry
Slovakia
ContactUsLinkComponent
Human Tau-441 (P301L), His Tag (TAU-H5144) is expressed from E.coli cells. It contains AA Met 1 - Leu 441 (Accession # P10636-8 (P30 1L)). 1 * 100 µG
Catalog # ACRBTAU-H5144100UG
Supplier:  ACRO Biosystems
Human Tau-441 (P301L), His Tag (TAU-H5144) is expressed from E.coli cells. It contains AA Met 1 - Leu 441 (Accession # P10636-8 (P30 1L)). 1 * 100 µG
Catalog # ACRBTAU-H5144100UG
Supplier:  ACRO Biosystems
Frequently Bought Together