Order Entry
Slovakia
ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (BA.1.1/Omicron) (SPN-C5224) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1(A67V, HV69-70del, T95I, G142D, VYY143-145del, N211del, L212I, ins214EPE, G339D, R 1 * 50 µG
Catalog # ACRBSPN-C5224-50UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike Trimer, His Tag (BA.1.1/Omicron) (SPN-C5224) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1(A67V, HV69-70del, T95I, G142D, VYY143-145del, N211del, L212I, ins214EPE, G339D, R 1 * 50 µG
Catalog # ACRBSPN-C5224-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together