Order Entry
Slovakia
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein, His Tag (BA.2*&BA.3*/Omicron) (NUN-C52Hu) is expressed from human 293 cells (HEK293). It contains A A Met 1 - Ala 419 (Accession # QHO62115.1(P13L, ERS31-33del, R203K, G204R, S413R)). The nucleocapsid mutations are id 1 * 100 µG
Catalog # ACRBNUN-C52HU100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein, His Tag (BA.2*&BA.3*/Omicron) (NUN-C52Hu) is expressed from human 293 cells (HEK293). It contains A A Met 1 - Ala 419 (Accession # QHO62115.1(P13L, ERS31-33del, R203K, G204R, S413R)). The nucleocapsid mutations are id 1 * 100 µG
Catalog # ACRBNUN-C52HU100UG
Supplier:  ACRO Biosystems
Frequently Bought Together