Order Entry
Slovakia
ContactUsLinkComponent
Human CD3 epsilon, Mouse IgG2a Fc Tag (CDN-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Acces sion # NP_000724.1). 1 * 100 µG
Catalog # ACRBCDN-H5253100UG
Supplier:  ACRO Biosystems
Human CD3 epsilon, Mouse IgG2a Fc Tag (CDN-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Acces sion # NP_000724.1). 1 * 100 µG
Catalog # ACRBCDN-H5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together