Order Entry
Slovakia
ContactUsLinkComponent
Mouse CD117, Fc Tag (CD7-M5259) is expressed from human 293 cells (HEK293). It contains AA Ser 25 - Thr 519 (Accession # P05532-2). 1 * 100 µG
Catalog # ACRBCD7-M5259100UG
Supplier:  ACRO Biosystems
Mouse CD117, Fc Tag (CD7-M5259) is expressed from human 293 cells (HEK293). It contains AA Ser 25 - Thr 519 (Accession # P05532-2). 1 * 100 µG
Catalog # ACRBCD7-M5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together