Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human LRP-6, His,Avitag (LR6-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Access ion # O75581-1 ). 1 * 200 µG
Catalog # ACRBLR6-H82E7200UG
Supplier:  ACRO Biosystems
Biotinylated Human LRP-6, His,Avitag (LR6-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Access ion # O75581-1 ). 1 * 200 µG
Catalog # ACRBLR6-H82E7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together