Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Cynomolgus CD79B, His,Avitag (CDB-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 161 (A ccession # A0A2K5WTH8-1). 1 * 200 µG
Catalog # ACRBCDB-C82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD79B, His,Avitag (CDB-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 161 (A ccession # A0A2K5WTH8-1). 1 * 200 µG
Catalog # ACRBCDB-C82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together