Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse Osteopontin, His,Avitag (OPN-M82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 17 - Asn 294 ( Accession # Q547B5-1). 1 * 200 µG
Catalog # ACRBOPN-M82E8200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse Osteopontin, His,Avitag (OPN-M82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 17 - Asn 294 ( Accession # Q547B5-1). 1 * 200 µG
Catalog # ACRBOPN-M82E8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together