Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human LRP-5, His,Avitag (LR5-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Acce ssion # O75197-1 ). 1 * 200 µG
Catalog # ACRBLR5-H82E6200UG
Supplier:  ACRO Biosystems
Biotinylated Human LRP-5, His,Avitag (LR5-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Acce ssion # O75197-1 ). 1 * 200 µG
Catalog # ACRBLR5-H82E6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together