Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus / Rhesus macaque IL-15, His Tag (IL5-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 ( Accession # P48092-1). 1 * 50 µG
Catalog # ACRBIL5-C52H4-50UG
Supplier:  ACRO Biosystems
Cynomolgus / Rhesus macaque IL-15, His Tag (IL5-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 ( Accession # P48092-1). 1 * 50 µG
Catalog # ACRBIL5-C52H4-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together