Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human OX40 Ligand, His,Avitag (OXL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 ( Accession # P23510-1). 1 * 200 µG
Catalog # ACRBOXL-H82Q5200UG
Supplier:  ACRO Biosystems
Biotinylated Human OX40 Ligand, His,Avitag (OXL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 ( Accession # P23510-1). 1 * 200 µG
Catalog # ACRBOXL-H82Q5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together