Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human FGF-7, His,Avitag (FG7-H82E7) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - Thr 194 (Access ion # P21781-1). 1 * 25 µG
Catalog # ACRBFG7-H82E7-25UG
Supplier:  ACRO Biosystems
Biotinylated Human FGF-7, His,Avitag (FG7-H82E7) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - Thr 194 (Access ion # P21781-1). 1 * 25 µG
Catalog # ACRBFG7-H82E7-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together