Order Entry
Slovakia
ContactUsLinkComponent
Mouse B7-H4, His Tag (B74-M52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gly 257 (Accession # Q7TSP5-1). 1 * 1 mg
Catalog # ACRBB74-M52H9-1MG
Supplier:  ACRO Biosystems
Mouse B7-H4, His Tag (B74-M52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gly 257 (Accession # Q7TSP5-1). 1 * 1 mg
Catalog # ACRBB74-M52H9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together