Order Entry
Slovakia
ContactUsLinkComponent
Human BST1, His Tag (ENN-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Gly 586 (Accession # P17813-1). 1 * 500 µG
Katalógové číslo ACRBENN-H52H9500UG
Supplier:  ACRO Biosystems
Human BST1, His Tag (ENN-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Gly 586 (Accession # P17813-1). 1 * 500 µG
Katalógové číslo ACRBENN-H52H9500UG
Supplier:  ACRO Biosystems
Frequently Bought Together